ContactUsLinkComponent
Microcentrifuge Tube Rack
Microcentrifuge Tube Rack
Catalog # 470149-162
Supplier:  Ward's Science
undefined
Microcentrifuge Tube Rack
Catalog # 470149-162
Supplier:  Ward's Science
Supplier Number:  241020
Specifications
  • Description:
    Microcentrifuge Tube Rack, 24 Place
  • Cat. no.:
    470149-162
  • Supplier No.:
    241020
Specifications
About this item
Economical and compact, this microtube rack comes pre-assmbeled and ready to hold up to twenty-four 1.5–2.0 ml microtubes. Its clearly labeled holes make sample identification easy, while its compact design saves benchspace and makes for easy storage.