Orders LinkContactUsLinkComponent
Metric Stacking Masses
Metric Stacking Masses
Metric Stacking Masses
Catalog # 470110-708
Supplier:   SI Manufacturing
Supplier #:  91630
Metric Stacking Masses
Catalog # 470110-708
Supplier:  SI Manufacturing
Metric Stacking Masses
Metric Stacking Masses
Metric Stacking Masses
Price
$28.45
Log in for availability
Kit of 52
Product Details & Documents
Designed to illustrate that height is proportional to mass, when diameter is uniform. Set consists of 52 identified masses which are molded to stack. Included are 4 yellow 10 g masses, eight red 5 g masses and 40 blue 1 g masses.
Specifications
  • Description:
    Metric Stacking Masses