Mega-Molecules Advanced Chemistry Molecular Kit
470120-980
- Description:Mega-Molecules Advanced Chemistry Molecular Kit
- Cat. no.:470120-980
- Supplier No.:MG155
The Advanced General and Organic Chemistry Molecular Model Set has all of the parts necessary to be successful in a college/university level General or Organic Chemistry course.
- Hands on activity
- Gain better understanding of molecular structure
This set contains 180 durable plastic parts including 5 VSEPR models (linear, trigonal planar, tetrahedral, trigonal bipyramidal, and octahedral) and 1 square planar model for building complex ions. The bond lengths are scaled for accuracy. This model set has both rigid and flexible bonds. Rigid bonds allow for easy rotations around single bonds. Flexible bonds are great for creating double and triple bonds and securely holding the atoms within the structures.