ContactUsLinkComponent
Aneroid Sphygmomanometer
Aneroid Sphygmomanometer
Catalog # 470019-064
Supplier:  HEALTH SMART SE
undefined
Aneroid Sphygmomanometer
Catalog # 470019-064
Supplier:  HEALTH SMART SE
Supplier Number:  01-100-011
Specifications
  • Description:
    Aneroid Sphygmomanometer
  • Cat. no.:
    470019-064
  • Supplier No.:
    01-100-011
Specifications
About this item
Features Markings On The Cuff For Better Accuracy
Pre-calibrated and self-zeroing, this model assures you consistently accurate measurements. It comes with a standard-size cuff with Velcro closure, instructions, and stores in its own leatherette case. Accurate to ±3 mm Hg.