Make Your Own Watershed Kit
Catalog # 470149-796
Supplier: JT & A, INC.
Specifications
- Description:Make your own Watershed Kit
- Cat. no.:470149-796
- Supplier No.:71005K-B
Specifications
About this item
Modeled after the award-winning EnviroScape models, this kit allows your students to create their own city and learn about water pollution at the same time.
- A great hands-on way to explore the environment
Instructional activities highlight how water pollution occurs and how to prevent it — such as adding cocoa for soil, drink mixes for chemicals, and making it rain. You can even make manure for use as a pollutant on the model – and experiment with techniques (best management practices) for preventing water pollution.
The kit includes plaster cast forms, three paint brushes, non-toxic acrylic paint, three felt strips, bridges, instructions, and activities.