Liver Slide
Liver Slide, Thin section, Hematoxylin and Eosin
Catalog # 470183-690
Supplier: Avantor
Jump to:
Product Details & Documents
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Routine preparation showing hepatic cells, Kupffer’s cells, blood vessels, and bile ducts.
Specifications
- Description:Liver Slide
- Stain:Hematoxylin and Eosin
- Orientation:Thin section