Orders LinkContactUsLinkComponent
Liter Cube
Liter Cube
Liter Cube
Catalog # 470014-884
Supplier:   SI Manufacturing
Supplier #:  77864
Liter Cube
Catalog # 470014-884
Supplier:  SI Manufacturing
Liter Cube
Liter Cube
Liter Cube
Price
$13.20
Log in for availability
Each
Product Details & Documents
Demonstrate the Relation of Length, Area, and Volume
  • Graduated in 100 mL intervals
  • Illustrate that volume = height x base
  • Convenient for measuring mass
This clear, acrylic liter cube is graduated to 1,000 mL in 100 mL intervals and has a cm grid on the bottom to illustrate that volume = height x base. The large top opening makes it convenient for activities measuring mass by the displacement of water.
Specifications
  • Description:
    Liter Cube