Liter Box
Liter Box
Catalog # 470112-470
Supplier: SI Manufacturing
Supplier #: 77864-A
Jump to:
Product Details & Documents
A perfect cubic decimeter box that contains exactly 1 liter. Can be used to demonstrate the metric relationships among linear, volume, one and mass measurements.
Specifications
- Description:Liter Box