Lister Bandage Scissors
470124-066
- Description:Lister Bandage Scissors
- Cat. no.:470124-066
- Supplier No.:25-702-000
Angled blades get underneath bandage layers.
- Stainless Steel Construction
- Ideal for Tightly Wrapped Surgical Bandages
- Clean, Accurate Cuts
The sharp, angled blades on these scissors allow them to maneuver behind tightly-wrapped surgical bandages for a clean and accurate cut.
The stainless steel scissors are 5½" long.