Lead(IV) Oxide
Lead(IV) Oxide, Laboratory, 100 g
: LL0137-100G
CAS Number: 1309-60-0
Formula Weight: 239.19
Formula: PbO2
Hazard Info: Toxic, Oxidizer
Density (g/mL): 9.2
Solubility: Acetic Acid
Synonyms: Lead Dioxide, Lead Peroxide
Shelf Life (months): 36
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
- Description:Lead(IV) Oxide
- Size:100 g
- Chemical/physical specifications:Powder
- Grade:Laboratory



