About this item
Easy to carry, simple to operate and sufficient for most field pH tests.
Highly stable and accurate readings with a large LCD display. Designed for simple one-hand operation. Automatically calibrates to pH 4, 7, 10 buffers and can be adjusted to recognize non-standard buffers as well. Features hold function, and auto power off with low battery indicator. The case is IP65 waterproof, and will float if accidentally dropped into the water.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 470204-168
- 470204-170
- pH range:
- 0 - 14
- 0 - 14
- pH resolution:
- 0.1
- 0.01
- pH accuracy:
- ±0.2
- ±0.05
- Dimensions:
- 165 × 38 × 38 mm (6.5 × 1.5 × 1.5")
- 165 × 38 × 38 mm (6.5 × 1.5 × 1.5")
- Weight:
- 2.5 oz (68 g)
- 2.5 oz (68 g)