If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:LaMotte® Plant Tissue Macronutrient Test Kit
- No. of Lab Groups:50
- Cat. no.:470006-954
- Supplier No.:5026-01
Specifications
About this item
What Soil Nutrients Do Your Plants Really Use?
- Select your own plant tissue extracts
- Convenient Carrying Case
- Includes a Soil Handbook
Compare plants from home, the field, or a hydroponic greenhouse to determine the amount of nutrients they receive from the soil. Just a single plant tissue extract can be used for colorimetric tests of nitrogen, potassium, and phosphorus (fifty tests per factor), and all results can be quantified by trace, low, medium, or high concentrations.
The kit also comes with material for students to prepare their own plant tissue extracts. A soil handbook and all reagents and labware are supplied in a sturdy plastic case.