Order Entry
Canada
ContactUsLinkComponent
Labnet Prism R Refrigerated Microcentrifuge
Labnet Prism R Refrigerated Microcentrifuge
Catalog # 470129-044
undefined
Labnet Prism R Refrigerated Microcentrifuge
Catalog # 470129-044
Supplier Number:  C2500-R

Specifications

  • Description:
    Labnet Prism R Refrigerated Microcentrifuge
  • Cat. no.:
    470129-044
  • Supplier No.:
    C2500-R

Specifications

About this item

This high performance, refrigerated centrifuge can cool to 4°C in just eight minutes.

  • Rotor Included
  • Refrigerated
  • Variable Speed: 13,500 rpm/17,135 x g.

The variable speed of this microcentrifuge is 13,500 rpm/17,135 x g.

It's supplied with a 24 x 1.5/2.0 mL rotor.