Lab Tools Kit
Kit, Lab Tools Set/6
Catalog # 470004-944
Supplier: United Scientific Supplies
Supplier #: LSET6
Jump to:
Product Details & Documents
Economical tool set for the lab.
- Test tube brush for cleaning glassware
- Tongs and tweezers for moving samples
Supply each student with their own integrated science six-piece tool kit.
Includes 13 cm tweezers, 13 cm test tube clamp, 23 cm crucible tongs, 18 cm test tube brush, 16.51 scoop with one pointed end and one blunt end, and 15.24 cm spatula/lifter and spoon.
All pieces except the test tube clamp are stainless steel.
Specifications
- Description:Kit, Lab Tools Set/6