Kick-Net
Kick-Net, 1 m, 1 m, 500 μm
Catalog # 470112-698
Supplier: LaMotte
Supplier #: 0021-P
Jump to:
Product Details & Documents
Easily Collect Benthic Macroinvertebrates
- GLOBE Approved
- Easily Transportable
Use this Kick-Net to perform USEPA Rapid Bioassessment Protocols on benthic macroinvertebrates. Net is one meter square of 500 micron material. Contains hemmed sides and reinforced bottom seams to ensure anchoring. Complete with poles.
Specifications
- Description:Kick-Net
- Length:1 m
- Mesh size:500 μm
- Width:1 m
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



