You searched for: Pasteur/Transfer Pipettes
Completely control the small quantity of liquid being manually dispensed with the Pasteur and transfer pipettes. The all-in-one devices are inert to most fluids for maximum working flexibility. Squeezing the small or large bulb, releases homogeneous droplets from the transparent, narrow stems that may come graduated for precise biological spotting or filling applications. Due to the challenge of properly washing the bulb, Pasteur and transfer pipettes are available in disposable as well as reusable models.
Polyethylene Transfer Pipettes
Supplier: VWR International
VWR* Transfer Pipette, with Extended Tip, Large Bulb Bulk, made of ultra-clear, LDPE, unbreakable, Sterile, eliminate contaminated rubber bulbs, broken glass and exposure to infectious materials, Drops: Precise 66 drops/ml, Heat selable suitable for LN2 -196C, Volume: 7ml
Expand 16 Items
VWR® Disposable Pasteur Pipettes, Glass, Plugged
Supplier: VWR International
These disposable glass pipettes are furnished with cotton plugs.
Expand 4 Items
VWR® Disposable Pasteur Pipettes, Glass
Supplier: VWR International
VWR® Disposable Pasteur Pipettes are manufactured from 3.3 expansion borosilicate glass, and feature fire-polished tops and a straight-cut tip.
Expand 3 Items
VWR® Disposable Pasteur Pipettes, Borosilicate Glass 7.0 Expansion
Supplier: VWR International
VWR® Disposable Pasteur Pipettes are ideally suited for use in both clinical and industrial laboratories.
Expand 3 Items
VWR® Disposable Transfer Pipettes
Supplier: VWR International
These unbreakable all-in-one pipettes eliminate the hazard of broken glass and exposure to infectious materials.
Expand 41 Items
VWR® Disposable Transfer Pipettes with Reference Lines
Supplier: VWR International
Pipettes are designed with a small tip to ensure consistent reproduction of the drop size.
Expand 2 Items
VWR® Disposable Transfer Pipettes, Graduated
Supplier: VWR International
VWR® Disposable Transfer Pipettes are ideal for quick and safe fluid transfer.
Expand 18 Items
Disposable Polyethylene Dropper Pipettes
Supplier: Avantor
Chemically inert polyethylene pipettes.
Expand 1 Items
Graduated Dropping Pipette
Supplier: Avantor
Made of heavy-duty flint glass with 1 ml graduations.
Expand 1 Items
Graduated Transfer Pipettes
Supplier: Avantor
Designed for consisent drop sizes.
Expand 3 Items
Disposable Polyethylene Dropper Pipettes
Supplier: Avantor
These polyethylene pipettes are chemically inert.
Expand 2 Items
KONTES® Disposable Pasteur Pipettes, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Single-use Pasteur pipettes for the transfer of small volumes.
Expand 2 Items
Corning® Gosselin™ Transfer Pipettes, Corning
Supplier: Corning
Corning® Gosselin™ offers a wide range of disposable sampling tools including Pipettes.
Expand 2 Items
SteriPettes, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
SteriPettes are sterile, disposable Pasteur pipettes produced from soda-lime, gamma-irradiated glass.
Expand 4 Items
Samco™ Extra Long Transfer Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Perform safe and easy sampling from large containers using Thermo Scientific™ Samco™ Extra-Long Transfer Pipettes.
Expand 6 Items
Kimble® Disposable Pasteur Pipettes, Soda Lime, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
These pipettes feature straight-cut tips and smooth, fire-polished tops to facilitate the attachment of bulbs.
Expand 1 Items
SP Bel-Art Transpette™ 8-Channel Transfer Pipet, Disposable, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Perform plate washing, media change, and supernatant collection without the use of pipettors and pipette tips with this inexpensive, 8-channel transfer pipet.
Expand 2 Items
Disposable Pasteur Pipettes, Borosilicate Glass, Wheaton
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
For use in inoculating culture media and as a general sampling pipet.
Expand 1 Items
Kimble Chase Glass Pasteur Pipettes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Pasteur Pipettes - Borosilicate and Flint Glass: Manufactured from the highest quality glass, these disposable pipettes feature uniform wall weight and tip diameters.
Expand 4 Items
WHEATON® Single-Use Polyethylene Pasteur Pipettes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Ideal for general liquid transfer applications and extracting samples for analysis.
Expand 3 Items
Plastic Transfer Pipettes, Argos Technologies
Supplier: Argos Technologies
Perfect for single use fluid transfers.
Expand 20 Items
Pipette, Disposable, Ace Glass Incorporated
Supplier: Ace Glass
Used for transferring blood in blood testing work and other areas where transfer of material is essential but accuracy not important.
Expand 2 Items
Nalgene® Dropper Pipette, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Molded in one piece construction.
Expand 1 Items
Plastic Dropper, Hach
Supplier: Hach
Plastic dropper with 0.5 and 1.0 ml mark (20 pack) is used in a variety of Hach test kits.
Expand 1 Items
Pasteur Pipettes, Argos Technologies
Supplier: Argos Technologies
Argos plastic pasteur pipettes are produced from medical-grade polystyrene in a class 100,000 cleanroom.
Expand 2 Items
Falcon® Disposable Transfer Pipettes, Corning
Supplier: Corning
PE, disposable.
Expand 2 Items
Samco™ Q-PET™ Disposable Transfer Pipet, Thermo Scientific
Supplier: Thermo Fisher Scientific
Nonsterile polyethylene transfer pipette designed to fit cuvettes.
Expand 1 Items
Volumetric Transfer Pipette
Supplier: Thermo Scientific
These pipettes are economical and convenient.
Expand 4 Items
Samco™ Sedi-Pet® Disposable Transfer Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Polyethylene pipettes are used for blood bank applications, such as sedimentation procedures.
Expand 6 Items
Samco™ Disposable Transfer Pipettes, Graduated, Thermo Scientific
Supplier: Thermo Fisher Scientific
Graduated polyethylene transfer pipettes are excellent for sampling of serum, plasma, blood, urine, and other biological fluids, especially where more accurate volume measurements are required.