Order Entry
Canada
ContactUsLinkComponent
Justrite® Liquid Disposal Can and ECO Funnels
Justrite® Liquid Disposal Can and ECO Funnels
  470202-914
 :  Justrite
undefined
Justrite® Liquid Disposal Can and ECO Funnels
  470202-914
 :  Justrite
 :  14765
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Capacity:
    5 gal. (19 L)
  • Description:
    HDPE Disposal Can
  • Environmentally Preferable:
  • Cat. no.:
    470202-914
  • O.D.×H:
    12×20"
  • Supplier No.:
    14765

 

 

A safe solution for containing flammable wastes and corrosives, this Justrite Liquid Disposal Can is constructed of high density polyethylene for superior chemical resistance and leak protection.

  • High-Density Polyethylene Construction
  • Flame Arrester Guards Against Outside Ignition
  • Innovative, FM-Approved Eco Funnels
  • Easy Pouring and Filling
  • Cap Vents at between Three and Five psig

The lid's fixed position on the lid allows for easy pouring and filling. When closed, the cap vents at between three and five psig while a flame arrester guards against outside ignition.

Designed to work with the Justrite container, innovative ECO funnels give users increased control and decreased chance of spills when pouring hazardous liquids. They feature a cap for safe sealing when not in use. All funnels are FM approved and equipped with a flame arrestor.

ECO Funnels