ContactUsLinkComponent
Infrared Thermometer with LED Light Pen
Infrared Thermometer with LED Light Pen
Catalog # 470204-106
Supplier:  Sper Scientific
undefined
Infrared Thermometer with LED Light Pen
Catalog # 470204-106
Supplier:  Sper Scientific
Supplier Number:  800100
Specifications
  • Description:
    IR Thermometer with LED Light Pen
  • Length:
    6½"
  • Cat. no.:
    470204-106
  • Range:
    –33…+220 °C (–27...428 °F)
  • Supplier No.:
    800100
Specifications
About this item
Collect Temperature Values with the Press of a Button.
  • Provides 35 hours of continuous use
  • Thermometer is combined with a bright LED
  • Ideal for field use
With a response time of one second, this non–contact infrared thermometer quickly and accurately scans for temperature in Celsius and Fahrenheit. The thermometer end is protected by a cap when not in use, while the other side of the pen features a super bright white LED. Especially useful in the field, this versatile tool can provide 35 hours of continuous temperature measurement. Includes one AAA battery, two L1154 button cell batteries, and an attractive foam lined metal case.