Introductory Muscle Tissues Slide Set
Catalog # 470183-578
Supplier: Avantor
Specifications
- Type:Histology
- Description:Introductory Muscle Tissues Slide Set
- No. of Slides:3
- Cat. no.:470183-578
- Supplier No.:WARD470183-578
Specifications
About this item
For Demonstration and Comparison of Types
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Smooth muscle; Cardiac muscle; Skeletal muscle. All are cross/longitudinal sections.