Introductory Connective Tissues Slide Set
Catalog # 470177-844
Supplier: Avantor
Specifications
- Type:Histology
- Description:Introductory Connective Tissues Slide Set
- No. of Slides:12
- Cat. no.:470177-844
- Supplier No.:WARD470177-844
Specifications
About this item
Survey of Vertebrate Connective Tissues
- Identifying features clearly distinguishable
- Rigorous quality control standards
All are sections unless otherwise specified.
Ordering information: Slide Set contents include: Mesenchyme, embryonic; Areolar (wm); Adipose, mesentery; Mucous, umbilical cord; White fibrous, tendon; Yellow elastic, ligamentum nuchae; Hyaline cartilage; Elastic cartilage, epiglottis; White fibro-cartilage, pubic symphysis; Osteogenesis, intramembranous ossification; Osteogenesis, endochondral ossification; Bone, ground preparation (thin section).