Orders LinkContactUsLinkComponent
Hook Collars
Hook Collars
Hook Collars
Certificates
Hook Collars
2 Options Available Below
Product Details & Documents
Quickly add a hook to a support rod.
  • Plated steel
  • Thumbscrew
Plated steel collar clamps feature thumbscrews and hooks for suspending pulleys, balances, etc. from support rods or crossbars.
Product Family Options
Product Information
  • Availability
    Price
  • 470006-700
    Hook Collar, Fits 13 mm (0.5") Rod
    Hook Collar, Fits 13 mm (0.5") Rod
    Log in for availability
    Log in for availability
    Each
    Each - $19.90
    $19.90
    Each
       $19.90
    Supplier Number: HKCL01
  • 470006-702
    Hook Collar, Fits 19 mm (0.75") Rod
    Hook Collar, Fits 19 mm (0.75") Rod
    Log in for availability
    Log in for availability
    Each
    Each - $21.95
    $21.95
    Each
       $21.95
    Supplier Number: HKCL02