Orders LinkContactUsLinkComponent
Heating Mantle Controller
Heating Mantle Controller
Heating Mantle Controller
Catalog # 470202-750
Supplier:   Glas-Col
Supplier #:  104A PL312
Heating Mantle Controller
Catalog # 470202-750
Supplier:  Glas-Col
Heating Mantle Controller
Heating Mantle Controller
Heating Mantle Controller
Price
$577.80
Log in for availability
Each
Product Details & Documents
Regulates Heating Mantles, Tapes and Cords, Small Furnaces, Ovens and Hot Plates.
  • Analog controller
  • Provides 5.5% to 100% full power
Anodized aluminum case with fuse, pilot light, and 6' grounded line cord. Size: 4¼" L x 2½" W x 2" H. For 120V, 50/60Hz, 1500W. CSA listed.
Specifications
  • Description:
    Heating Mantle Controller