Orders LinkContactUsLinkComponent
Hardness Points and Testing Set
Hardness Points and Testing Set
Hardness Points and Testing Set
Catalog # 470004-368
Supplier:   MINERALAB, LLC
Supplier #:  HD1
Hardness Points and Testing Set
Catalog # 470004-368
Supplier:  MINERALAB, LLC
Hardness Points and Testing Set
Hardness Points and Testing Set
Hardness Points and Testing Set
Price
$342.75
Log in for availability
Kit of 1
Product Details & Documents
Identify Minerals with This Convenient Kit
This unique set contains three mineral tests in one box, plus the innovation of the hardness testing points. Unlike other tools that chip and break, the four double-ended picks in this set are made of durable ground metal and alloy points. The points can be resharpened easily with the included grinding stone. For ultimate convenience, the picks are stamped with the indicated hardness value, 2 through 9 on Mohs’ hardness scale, and are color-coded for quick identification. Packaged in one neatly arranged, compact wooden case, the set also includes a streak plate, glass plate, and instructions.
Specifications
  • Description:
    Hardness Points and Testing Set