ContactUsLinkComponent
GREEN Low Cost Estuary and Marine Water Monitoring Kit
GREEN Low Cost Estuary and Marine Water Monitoring Kit
Catalog # 470146-202
Supplier:  LaMotte
undefined
GREEN Low Cost Estuary and Marine Water Monitoring Kit
Catalog # 470146-202
Supplier:  LaMotte
Supplier Number:  5911
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    LaMotte GREEN Low Cost Estuary & Marine Water Monitoring Kit
  • No. of Lab Groups:
    10
  • Cat. no.:
    470146-202
  • Supplier No.:
    5911
Specifications
About this item
Economical Water Testing Solution
  • No hazardous compounds
  • Can be used in the field or the lab
  • Single factor tests
Designed to provide simple, economical, and non-hazardous methods for testing saline and brackish water, this kit provides 10 tests each for pH, DO, BOD, temperature, turbidity, phosphate, chloride, and two for total coliform. All contents and chemicals are non-hazardous and field-friendly.