Glass Buret Funnell, Montclare Scientific
470120-586
- Description:Glass Buret Funnel
- Cat. no.:470120-586
- Supplier No.:QUOTE 2-93
For filling burets. Diameter: 35 mm; total length: 60 mm; outside diameter of stem: 6 mm.
For filling burets. Diameter: 35 mm; total length: 60 mm; outside diameter of stem: 6 mm.