Gilley Induction Coil Set
Catalog # 470005-394
Supplier: Wards
Specifications
- Description:Gilley Induction Coil Set
- Environmentally Preferable:
- Cat. no.:470005-394
- Supplier No.:GLCL
Specifications
About this item
An excellent set of materials for demonstrating electromagnetism and induced currents, the set includes two electromagnetic coils mounted on plastic saddle blocks, round and square iron cores, split round iron cores, a double core, and a card for showing lines of force. Set of two.