General Bacteriology Slide Set
General Bacteriology Slide Set, 10, Monera
Catalog # 470183-434
Supplier: Avantor
Jump to:
Product Details & Documents
A Review of the Basic Form and Structure of Bacteria
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Bacteria forms, separate smears; Azotobacter chroococcum; Agrobacterium tumefaciens; Bacillus anthracis; Bacillus subtilis; Lactococcus lactis; Mycobacterium sp.; Proteus hauseri; Rhodospirillum rubrum; and Streptococcus pyogenes.
Specifications
- Type:Monera
- Description:General Bacteriology Slide Set
- No. of slides:10