Ganoid, Scale Slide
Ganoid, Scale, Whole mount, Schneider's
470181-716
: Avantor
Fish Scales that do not Overlap
- Identifying features clearly distinguishable
- Rigorous quality control standards
Stained, isolated scale of bony structure. These scales are represented on gars, sturgeon, and bowfin, among others.
- Description:Ganoid, Scale
- Stain:Schneider's
- Orientation:Whole mount