Orders LinkContactUsLinkComponent
Fundamentals of Physics Reflection/Refraction Apparatus
Fundamentals of Physics Reflection/Refraction Apparatus
FUNdamentals of Physics Reflection/Refraction Apparatus, 1
Catalog # 470014-332
Supplier:   Wards
Supplier #:  RRKT
Fundamentals of Physics Reflection/Refraction Apparatus
Catalog # 470014-332
Supplier:  Wards
Fundamentals of Physics Reflection/Refraction Apparatus
Fundamentals of Physics Reflection/Refraction Apparatus
FUNdamentals of Physics Reflection/Refraction Apparatus, 1
Price
$180.80
Log in for availability
Each
Product Details & Documents
Hands-on investigation even explores light separation.
  • 360 degree protractor
  • Laser and light source
  • Includes teacher manual
Using a plane mirror in addition to a concave and convex mirror with different focal lengths, students encounter reflection through a series of activities. Students also investigate refraction of concave and convex lenses, a Lucite block, and glass block. The base has a 12 V light bulb for investigating light dispersion and also a laser pointer for refraction. The included prism is utilized to demonstrate separation of light into its component colors. Comes with an instruction manual. Required but not included is a 12 V power supply.
Specifications
  • Description:
    FUNdamentals of Physics Reflection/Refraction Apparatus
  • No. of lab groups:
    1