Orders LinkContactUsLinkComponent
Functional Lung Model
Functional Lung Model
Supplier:   GSC International
Functional Lung Model
Supplier:  GSC International
Certificates
Functional Lung Model
Functional Lung Model
1 Options Available Below
Product Details & Documents
An Inexpensive Demonstration of Respiration.
  • Easy to Assemble
  • Includes All Required Parts
This unique and easy-to-use model features parts that replicate the actions of respiration.
Pulling on the rubber diaphragm that covers the bell jar causes the balloon lungs to fill with air as air pressure in the jar is reduced. When users push the diaphragm in, the balloons collapse and recreate exhalation.
Ordering information: The set includes an acrylic bell jar, Y tube, two balloons, a rubber diaphragm with a knob for ease of movement, and an instruction manual.
Product Family Options
Product Information
  • Width
    Length
    Height
    Availability
    Price
  • 470235-694
    Functional Lung
    Functional Lung
    19 cm
    19 cm
    19 cm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $163.05
    $163.05
    Log in for availability
    Case of 8
    Case of 8 - $1,274.10
    $1,274.10
    Each
       $163.05
    Case of 8
       $1,274.10
    Supplier Number: 120310-8
  • Discontinued
    Supplier Number: -