Orders LinkContactUsLinkComponent
Full Spear Point Dissecting Needle
Full Spear Point Dissecting Needle
Full Spear Point Dissecting Needle, Stainless steel, 152 mm (6"), Spear
Catalog # 470018-970
Supplier:   Dunrite Instruments
Supplier #:  141035
Full Spear Point Dissecting Needle
Catalog # 470018-970
Full Spear Point Dissecting Needle
Full Spear Point Dissecting Needle, Stainless steel, 152 mm (6"), Spear
Price
$9.25
Log in for availability
Each Box of 50
Product Details & Documents
The spear point is useful for cutting fascia and cleaning tissue from small structures.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Description:
    Full Spear Point Dissecting Needle
  • Length:
    152 mm (6")
  • Material:
    Stainless steel
  • Tip style:
    Spear