ContactUsLinkComponent
Ward's® Fruit DNA Extraction Activity Lab Activity
Ward's® Fruit DNA Extraction Activity Lab Activity
Catalog # 470013-266
Supplier:  Avantor
undefined
Ward's® Fruit DNA Extraction Activity Lab Activity
Catalog # 470013-266
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Fruit DNA Extraction Activity Kit
  • No. of Lab Groups:
    40
  • Cat. no.:
    470013-266
  • Supplier No.:
    WARD470013-266
Specifications
About this item
A low-tech solution for teaching advanced biotechnology.
  • Materials for 40 DNA extractions
  • Time required: 45 minutes
Demonstrate the physical properties of DNA with this comprehensive kit featuring all the necessary materials for completing fruit DNA extraction and analysis. All you add is the fruit. Specially prepared cell lysis solution and DNA precipitating solution allow students to easily and quickly extract DNA by first crushing their fruit, lysing the cell membrane to release the cell components, and finally precipitating the DNA. The students will then view the DNA by spooling it onto a plastic loop. This process illustrates the enormous length of the DNA molecule and provides a visualization of DNA that will help students understand its physical properties. Kit contains step-by-step student instructions, and teacher background information.