ContactUsLinkComponent
Friction Block Set
Friction Block Set
Catalog # 470149-098
undefined
Friction Block Set
Catalog # 470149-098
Supplier Number:  PH10580A
Specifications
  • Description:
    Friction Block Set
  • Cat. no.:
    470149-098
  • Supplier No.:
    PH10580A
Specifications
About this item
The Friction Block Set is ideal for simple friction experiments.
  • Ideal for Simple Friction Experiments
  • Instructions Included
  • Attached Mirror
This set consists of a glass mirror with sandpaper affixed to one side and two wooden blocks: one with a hook and the other with an eye. The latter block also has a mirror cemented to one face.
It comes with instructions for use.