Freefall Apparatus
Product Details & Documents
New Design for More Accuracy
Students can observe and measure the acceleration of a falling object. The unique drop and catch design, when used with the Fun Stand, allows an object to fall directly through a pair of photogates. The exact value for gravity can then be calculated for your local geographic area. The two included marbles have different masses, so the experiment can be repeated mutliple times to illustrate the universal effect of gravity.
Ordering information: Requires the Fun Stand and Photogate System. Includes a wooden dropping device, catching device, two marbles of different mass, two mounting screws, instructions, and activity guide.
Caution: WARNING: CHOKING HAZARD–Small Parts. Not for children under 3 yrs.
Specifications
- Description:FUNdamentals of Physics Free Fall Apparatus
- Height:2"
- Length:4"
- No. of lab groups:1
- Width:4"
Accessories
Product Information
- Catalog #DimensionsWeightAvailabilityPrice
- Fundamentals of physics photogate systemFundamentals of physics photogate system470014-46616×10×6 cm0.5 kg (1.1 lbs.)Log in for availabilityEach - $530.35
Product Information
- Catalog #DimensionsAvailabilityPrice
- Fundamentals of physics fun stand
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



