Four-Channel Timer
470001-920
- Description:Four-channel timer
- Cat. no.:470001-920
- Supplier No.:TI388
This timer features a clock and four independent channels that count up or down from any interval from one second to 100 hours — each with its own distinctive alarm.
It will automatically count up after the countdown reaches zero. It features a stand, pocket clip, magnet, and a quartz crystal accurate clock display. It runs on a 1.5 V battery, included. Size: 2⁷/₈L×2¹/₂W×⁹/₁₆"H.