United Scientific™ Push/Pull Spring Scales
Versatile spring scale.
- Weighs in Grams and Newtons
- Color coded to differentiate capacity
These Push Pull Spring Scales are a versatile spring scale that can be used to measure forces using push or pull functions, or to weigh a suspended object from the large hook.
: Includes a zero adjustment screw, a metal suspension ring, instructions and application sheets.
: This product is for ages six and up.
Frequently Bought Together