Folding Pocket Magnifier
Catalog #: 470144-826
Supplier: United Scientific Supplies
Specifications
- Magnification:2.5×
- Description:Folding Pocket Magnifier
- Cat. no.:470144-826
- Supplier No.:MPF050
Specifications
About this item
This compact magnifier includes a hinged leather cover.
- Magnification: 2.5x
- Diameter: 5 cm
- Glass Construction
- Hinged Leather Cover Provided
The magnifier is 5 cm in diameter, and its magnification is 2x.
The lens is made of glass for clarity.
Customers Who Bought This Also Bought