Order Entry
Canada
ContactUsLinkComponent
Fluorescent Mineral Collection with UV Lamp
Fluorescent Mineral Collection with UV Lamp
Catalog # 470095-642
Supplier:  POLMAN MINERALS
undefined
Fluorescent Mineral Collection with UV Lamp
Catalog # 470095-642
Supplier:  POLMAN MINERALS
Supplier Number:  30310-06

Specifications

  • Description:
    Fluorescent Mineral Collection with UV Lamp
  • No. of Samples:
    10
  • Cat. no.:
    470095-642
  • Supplier No.:
    30310-06

Specifications

About this item

Dive into the colorful world of fluorescent minerals with this all-in-one educational set.

You'll be fascinated as you witness the dramatic color changes in the 10 carefully chosen fluorescent mineral samples viewed under the light of the included powerful longwave LED flashlight.

The mineral samples average 1" x 1" in size and are permanently mounted and labeled in a specially-designed display box measuring 8 1/2" x 9" x 2".

The LED flashlight stores conveniently in the box with two clamps and requires three AAA batteries to operate (not included).