Field Soil Test Kit
Product Details & Documents
Illustrate the Chemical Variety in Soil
- Provides accurate and informative results
- Includes recommendations for fertilizer use
- Includes a teacher's guide
Students will test for nitrogen, phosphorus, potassium, and pH using syringes and test tubes to obtain accurate results. The supplied reference cards give information about each test, and offer recommendations for applying fertilizer. The kit contains enough reagents and materials for 120 tests and a teacher’s guide and comes in a sturdy carrying case.
Specifications
Specifications
- Description:Field Soil Test Kit
- No. of tests:120
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.