ContactUsLinkComponent
Feathers, Composite Slide
Feathers, Composite Slide
Catalog # 470176-950
Supplier:  Avantor
undefined
Feathers, Composite Slide
Catalog # 470176-950
Supplier:  Avantor
Specifications
  • Description:
    Feathers, Composite
  • Cat. no.:
    470176-950
  • Stain:
    Composite: Multiple stains
  • Supplier No.:
    WARD470176-950
  • Orientation:
    Whole mount
Specifications
About this item
Compare and Contrast Feather Structure and Function
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Three types: contour, down, and filoplume.