ContactUsLinkComponent
Fast-Drying Epoxy Mounting Medium
Catalog # 470020-092
Supplier:  ELLSWORTH ADHESIVE
CAS Number:  
Fast-Drying Epoxy Mounting Medium
Catalog # 470020-092
Supplier:  ELLSWORTH ADHESIVE
Supplier Number:  608
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Fast-Drying Epoxy Mounting Medium
  • Cat. no.:
    470020-092
  • Supplier No.:
    608
Specifications
About this item
Process Slides in Just One Day
For fast results, mix the hardener (1.4 oz.) and resin (1.4 oz.), mount the chip to the slide, and the section will set in five minutes! Curing takes just 1–2 hours.