Order Entry
Canada
Orders LinkContactUsLinkComponent
<i>Eurypterus remipes</i>

Eurypterus remipes

Eurypterus remipes replica, 5", 7"
Catalog # 470226-644
Supplier:   Avantor
<i>Eurypterus remipes</i>
Catalog # 470226-644
Supplier:  Avantor
<i>Eurypterus remipes</i>
Eurypterus remipes

Eurypterus remipes replica, 5", 7"

Price
$112.95
Log in for availability
Each
Product Details & Documents

A complete and highly detailed "Sea scorpion", New York State's official fossil.

  • Silurian
  • Resin reproduction
  • New York
Specifications
  • Width:
    7"
  • Description:
    Eurypterus remipes replica
  • Length:
    5"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.