Euglena Model
470002-686
- Description:Euglena
- Height:15 cm
- Length:44 cm
- Cat. no.:470002-686
- Width:15 cm
- Supplier No.:B-147
- No. of Parts:1
Remarkable View Of An Autotrophic Protist
View the fascinating structures of a creature that exhibits the characteristics of both plants and animals. The one-piece model, enlarged 9,000X, is sectioned to show the interior of the cell, including the flagellum, stigma, blepharoplast, contractile vacuole, chloroplast, pyrenoids, rhizoplast, and endosome. It is mounted on a base and comes with a key, identifying 19 structures.