Orders LinkContactUsLinkComponent
Euglena
Euglena
Euglena, Whole Mount, Schneiders and Fast Green
Catalog # 470177-496
Supplier:   Avantor
Euglena
Catalog # 470177-496
Supplier:  Avantor
Euglena
Euglena
Euglena, Whole Mount, Schneiders and Fast Green
Price
$8.70
Log in for availability
Each
Product Details & Documents
Explore this often studied flagellate
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
General study. Demonstrates nucleus, chloroplasts, contractile vacuole, and flagellum.
Specifications
  • Description:
    Euglena
  • Stain:
    Schneiders and Fast Green
  • Orientation:
    Whole Mount
Accessories
Product Information
  • Catalog #
    Media
    No. of Students
    Availability
    Price
  • Euglena sp. Study Set
    Euglena sp. Study Set
    470189-298
    Euglena medium
    30
    Log in for availability
    Kit of 1 - $30.00
Product Information
  • Catalog #
    Media
    No. of Students
    Availability
    Price
  • Euglena sp (Large)
    Euglena sp (Large)
    470180-112
    Euglena medium
    100
    Log in for availability
    Each - $47.30
Product Information
  • Catalog #
    Media
    No. of Students
    Availability
    Price
  • Euglena gracilis (Small)
    Euglena gracilis (Small)
    470175-818
    Euglena medium
    30
    Log in for availability
    Each - $15.70