Orders LinkContactUsLinkComponent
Eudorina
Eudorina
Eudorina (Small), Basic culture solution, 30, Tube
Catalog # 470176-630
Supplier:   Ward's Science
Supplier #:  161760
Eudorina
Catalog # 470176-630
Supplier:  Ward's Science
Eudorina
Eudorina
Eudorina (Small), Basic culture solution, 30, Tube
Price
15,70 $
Log in for availability
Each
Product Details & Documents
Freshwater. Colonial; with spherical, biflagellated cells. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Specifications
  • Description:
    Eudorina (Small)
  • No. of students:
    30
  • Packaging:
    Tube
  • Media:
    Basic culture solution