Esophagus Slide
Esophagus Slide, Thin section, Hematoxylin and Eosin
Catalog # 470183-682
Supplier: Avantor
Jump to:
Product Details & Documents
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Stratified squamous epithelium, glands of submucosa, and two layers of muscularis are clearly shown.
Specifications
- Description:Esophagus Slide
- Stain:Hematoxylin and Eosin
- Orientation:Thin section