Order Entry
Canada
Orders LinkContactUsLinkComponent
Erlenmeyer Flask Assortment

Erlenmeyer Flask Assortment

Flask Set, 50 / 125 / 250 / 500 / 1000 ml
Catalog # 470122-272
Supplier #:  FGSET5
Erlenmeyer Flask Assortment
Catalog # 470122-272
Erlenmeyer Flask Assortment
Erlenmeyer Flask Assortment

Flask Set, 50 / 125 / 250 / 500 / 1000 ml

Price
$147.90
Log in for availability
Case of 1
Product Details & Documents

Includes the Five Most Popular Sizes.

  • Ideal for start up labs

Borosilicate glass flasks are designed to give the best combination of thermal shock resistance and physical strength. Feature heavy-duty rims and graduations in durable white enamel.

Ordering information: Set includes one of each of the following: 50 mL, 125 mL, 250 mL, 500 mL, 1000 mL.

Specifications
  • Capacity:
    50 / 125 / 250 / 500 / 1000 ml
  • Description:
    Flask Set