Ward's® Erdschreiber's Medium
Catalog # 470180-760
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Bottle
- Description:Erdschreiber’s Medium
- Size:1 L
- Cat. no.:470180-760
- Supplier No.:WARD470180-760
Specifications
About this item
Erdschreiber's Medium for culture of marine algae
- Sterile- no autoclaving
- Convenient - prepared for you
- Prepared consistenly every time
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.