Orders LinkContactUsLinkComponent
Equilateral Prisms
Equilateral Prisms
Prism, Equilateral, 25 mm x 50 mm
Catalog # 470003-426
Supplier #:  PGE050
Equilateral Prisms
Catalog # 470003-426
Equilateral Prisms
Equilateral Prisms
Prism, Equilateral, 25 mm x 50 mm
Price
$24.65
Log in for availability
Each
Product Details & Documents
Easily demonstrate the separation of light.
  • Demonstrate reflection and refraction
  • Flint glass
Excellent quality, polished flint glass prisms for experiments in diffraction, dispersion, and spectrum formation. Individually boxed to prevent chipping.
Specifications
  • Description:
    Prism, Equilateral, 25 mm x 50 mm