Specifications
- Description:Enhanced Student Spectrometer
- Cat. no.:470110-202
- Supplier No.:SPECT02
Specifications
About this item
Many advanced features in this economical instrument aid student learning in this Economical Instrument.
Many Advanced Features Aid Student Use in this Economical InstrumentOur Enhanced Student Spectrometer is a solidly-constructed instrument, thoughtfully provided with features and accessories that assist students in making precise measurements of spectral lines or prism angles. Mounted on a heavy cast-iron triped base, the spectrometer has excellent bearings for smooth, precise operation of the telescope, table, and divided circle and a 5mm slit mechanism. The instrument comes with a prism, a 300 lines/mm replica grating and holder, an autocollimation mirror, a power supply, adjustment tools, and a full instruction manual. Size: base spread 24cm, overall height 26cm.Built-in features include:• An illuminated autocollimation system for easy, rapid setup to ensure correct optical alignment of the instrument• A 2.2cm diameter achromatic optical system with a helical focuser for eyepiece accommodation to give clear, sharp images• A 17.8cm diameter, 360° x 0.5° divided circle with clear graduations and a double-ended inside vernier 0–30' x 1', equipped with an illuminated magnifier for easy reading. Circle and verniers are fully rotatable and have 4° range fine adjustments engaged by lockscrews. • A 7cm diameter prism and grating table, fully rotatable, with leveling screws, a prism holder, an engraved ring/radius pattern, and a 2cm height adjustment to accommodate any size of prism or grating
- Illuminated Auto-Collimation System
- Instructions
- Prism
- Auto-Collimation Mirror
- Cast-Iron Tripod Base
Our Enhanced Student Spectrometer is a solidly-constructed instrument provided with features and accessories that assist students in making precise measurements of spectral lines or prism angles.
Mounted on a heavy cast-iron tripod base, the spectrometer has excellent bearings for smooth, precise operation of the telescope, table, a divided circle, and a 5 mm slit mechanism. It also features an illuminated auto-collimation system for easy, rapid setup to ensure correct optical alignment of the instrument. Circle and verniers are fully rotatable and have 4° range fine adjustments engaged by lock screws.